Learn More
Medchemexpress LLC HY-P1260A 5mg Medchemexpress, FSLLRY-NH2 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1260A5MG
Encompass_Preferred
Medchemexpress, HY-P1260A 5mg FSLLRY-NH2 (TFA) CAS: FSLLRY-NH2 TFA is a protease-activated receptor 2 (PAR2) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3046