Learn More
Cayman Chemical FSLLRY-amide, Purity: ≥95%, CAS: 245329-02-6, 10 mg
SDP
Catalog No. 502971726
Encompass_Preferred
A peptide antagonist of PAR2 which blocks activation by trypsin (IC50 = ~50 μM) but not by the synthetic agonist SLIGRL-NH2; does not affect thrombin-induced activation of PAR1 or trypsin-mediated proteolysis of casein; used to study PAR2 action in cells and in animals
Catalog No. 50-297-1726 Supplier Cayman Chemical Supplier No. 1470910 MG
A peptide antagonist of PAR2 which blocks activation by trypsin (IC50 = ~50 μM) but not by the synthetic agonist SLIGRL-NH2; does not affect thrombin-induced activation of PAR1 or trypsin-mediated proteolysis of casein; used to study PAR2 action in cells and in animals