Learn More
Cayman Chemical FSlRY-amIde 1mg
SDP
Catalog No. 502971231
Encompass_Preferred
A peptide antagonist of PAR2 which blocks activation by trypsin (IC50 = ~50 μM) but not by the synthetic agonist SLIGRL-NH2; does not affect thrombin-induced activation of PAR1 or trypsin-mediated proteolysis of casein; used to study PAR2 action in cells and in animals
Catalog No. 50-297-1231 Supplier Cayman Chemical Supplier No. 147091 MG
A peptide antagonist of PAR2 which blocks activation by trypsin (IC50 = ~50 μM) but not by the synthetic agonist SLIGRL-NH2; does not affect thrombin-induced activation of PAR1 or trypsin-mediated proteolysis of casein; used to study PAR2 action in cells and in animals