Learn More
Cayman Chemical FSP1 Enzyme, 1 ea
SDP
Catalog No. 502896176
Encompass_Preferred
FSP1 Enzyme has been tested and formulated to work exclusively with Cayman’s FSP1 Fluorescent Inhibitor Screening Assay Kit (Item No. 701900).
Catalog No. 50-289-6176 Supplier Cayman Chemical Supplier No. 7019021 EA
FSP1 Enzyme has been tested and formulated to work exclusively with Cayman’s FSP1 Fluorescent Inhibitor Screening Assay Kit (Item No. 701900).