Learn More
Cayman Chemical FSP1 FluorScent InhIbItr Sc
SDP
Catalog No. 502892283
Encompass_Preferred
Cayman's FSP1 Fluorescent Inhibitor Screening Assay Kit provides a robust and EAsy-to-use platform for identifying novel inhibitors of human FSP1, a negative regulator of the ferroptosis pathway. The assay measures the reduction of resazurin by FSP1 into resorufin, which can be EAsily quantified using a fluorescence plate reader at excitation and emission wavelengths of 540 and 590 nm, respectively. The potent and reversible FSP1 inhibitor iFSP1 is included as a positive control and inhibits FSP1 with an IC50 value of 267 ± 47 nM.
Catalog No. 50-289-2283 Supplier Cayman Chemical Supplier No. 701900384 WELL
Cayman's FSP1 Fluorescent Inhibitor Screening Assay Kit provides a robust and EAsy-to-use platform for identifying novel inhibitors of human FSP1, a negative regulator of the ferroptosis pathway. The assay measures the reduction of resazurin by FSP1 into resorufin, which can be EAsily quantified using a fluorescence plate reader at excitation and emission wavelengths of 540 and 590 nm, respectively. The potent and reversible FSP1 inhibitor iFSP1 is included as a positive control and inhibits FSP1 with an IC50 value of 267 ± 47 nM.