Learn More
New England Biolabs, Inc. FspI – 2500 units
SDP
Catalog No. 50811992
Encompass
A restriction endonuclease that recognizes the sequence TGC_^GCA.
Catalog No. 50-811-992 Supplier New England Biolabs, Inc. Supplier No. R0135L
A restriction endonuclease that recognizes the sequence TGC_^GCA.