Learn More
Aviva Systems Biology Corp FST Antibody - C-terminal region (ARP42287_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42287_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FST. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FST. Alternate names: FSApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-58173