Learn More
Fine Science Tools Fine Scissors - Sharp, 9cm
SDP
Catalog No. NC9639443
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 22mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: No
Catalog No. NC9639443 Supplier Fine Science Tools Supplier No. 1406109
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 22mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: No