Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP542820UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Untagged. Host: E. coli. Species Reactivity: Mouse. BioActivity: Mouse Follistatin / FST induces MCP-11 cell proliferation (typical ED50 ? 1 ug/ml). AA Sequence: MGNCWLRQAK NGRCQVLYKT ELSKEECCST GRLSTSWTEE DVNDNTLFKW MIFNGGAPNC IPCKETCENV DCGPGKKCRM NKKNKPRCVC APDCSNITWK GPVCGLDGKT YRNECALLKA RCKEQPELEV QYQGKCKKTC RDVFCPGSST CVVDQTNNAY CVTCNRICPE PSSSEQYLCG NDGVTYSSAC HLRKATCLLG RSIGLAYEGK CIKAKSCEDI QCGGGKKCLW DSKVGRGRCS LCDELCPDSK SDEPVCASDN ATYASECAMK EAACSSGVLL EVKHSGSCN. Buffer: 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5. Purity: Greater than 90 as determined by SDS-PAGE. Endotoxin: Less than or equal to 5.00 EUs/ug as determined by the LAL method. Gene Symbol: FST. Gene ID: 14313. Uniprot: P47931.
Catalog No. 50-188-5828