Learn More
Fine Science Tools Semken Forceps, Stainless Steel, Straight, 15 cm
SDP
Catalog No. NC9046020
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 1.5 x 1.1mm/ Alloy / Material: Stainless Steel/ Length: 15cm
Catalog No. NC9046020 Supplier Fine Science Tools Supplier No. 1100815
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 1.5 x 1.1mm/ Alloy / Material: Stainless Steel/ Length: 15cm