Learn More
Medchemexpress LLC FStAx-35R TFA | 97.4% | 2885.36 (free base) | 1 MG
SDP
Catalog No. 5000426302
Encompass_Preferred
fStAx-35R TFA is a hydrocarbon-stapled peptide that inhibits the Wnt/β-catenin signaling pathway by disrupting the β-catenin-TCF interaction. It can be utilized in cancer research.
  • Inhibits Wnt/β-catenin signaling pathway
  • Disrupts β-catenin-TCF interaction
  • Suitable for cancer research
Catalog No. 50-004-26302 Supplier Medchemexpress LLC Supplier No. HYP10392AF1MG
fStAx-35R TFA is a hydrocarbon-stapled peptide that inhibits the Wnt/β-catenin signaling pathway by disrupting the β-catenin-TCF interaction. It can be utilized in cancer research.
  • Inhibits Wnt/β-catenin signaling pathway
  • Disrupts β-catenin-TCF interaction
  • Suitable for cancer research