Learn More
Aviva Systems Biology Corp FSTL5 Antibody - middle region (ARP57353_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57353_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FSTL5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FSTL5. Alternate names: DKFZp566D234, KIAA1263ApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53942