Learn More
Apexbio Technology LLC FT-207 (NSC 148958) 17902-23-7 100mg
SDP
Catalog No. 502240768
Encompass_Preferred
FT-207 (NSC 148958 CAS 17902-23-7) also known as tegafur or ftorafur is an orally administered prodrug of 5-fluorouracil (5-FU) Upon metabolic activation FT-207 releases 5-FU which inhibits thymidylate synthase and disrupts DNA synthesis thereby exhibiting cytotoxic effects on proliferating cells FT-207 is utilized in oncology research as a representative agent for studying fluoropyrimidine-based chemotherapy including combination regimens such as tegafur-uracil Its pharmacokinetic profile and conversion mechanism make it relevant for investigating prodrug strategies and anticancer therapeutics in preclinical models
This item is not returnable. View return policy
Catalog No. 50-224-0768 Supplier Apexbio Technology LLC Supplier No. B1474100
FT-207 (NSC 148958 CAS 17902-23-7) also known as tegafur or ftorafur is an orally administered prodrug of 5-fluorouracil (5-FU) Upon metabolic activation FT-207 releases 5-FU which inhibits thymidylate synthase and disrupts DNA synthesis thereby exhibiting cytotoxic effects on proliferating cells FT-207 is utilized in oncology research as a representative agent for studying fluoropyrimidine-based chemotherapy including combination regimens such as tegafur-uracil Its pharmacokinetic profile and conversion mechanism make it relevant for investigating prodrug strategies and anticancer therapeutics in preclinical models