Learn More
Medchemexpress LLC HY-111551 5mg Medchemexpress, FT113 CAS:1630808-89-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1115515MG
Encompass_Preferred
Medchemexpress, HY-111551 5mg FT113 CAS:1630808-89-7 FT113 is a potent and orally active fatty acid synthase (FASN) inhibitor, with an IC 50 of 213 nM for full-length recombinant human FASN enzyme. In cell-based assay, FT113 blocks FASN activity in BT474 cells (IC 50 , 90 nM). FT113 shows anti-proliferative activity, and exhibits anti-cancer activity both in vitro and in vivo [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2373