Learn More
Crystal Chem Inc Free Triiodothyronine (fT3) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80986
Encompass
The Free Triiodothyronine ELISA is used to quantify levels of human free T3 in serum using only a 25 µL sample volume. Highlights: Kits use sample size of only 25 µL. High sensitivity of 0.3 pg/mL. Dynamic range of 1 - 40 pg/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7887