Learn More
Medchemexpress LLC HY-112285 5mg Medchemexpress, FT895 CAS:2225728-57-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1122855MG
Encompass_Preferred
Medchemexpress, HY-112285 5mg FT895 CAS:2225728-57-2 FT895 is a potent and selective HDAC11 inhibitor with an IC50 of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9721