Learn More
Julabo USA Inc FT900 IMMERSION COOLER-90+30C; 300W, 115V/60Hz
SDP
Catalog No. 507533999
Encompass_Preferred
Chillers, Julabo, FT900 Immersion Cooler -90¿+30deg.C; 300W cooling power. JULABO Immersion Coolers provide counter-cooling when connected to a heating circulator or rapidly cool fluids down to a low temperature. With easy operation the units are suitable for a variety of cooling applications. Units are an alternative to cooling with dry ice or tap water.The FT200, FT400 and F900 units simply turn on and the probe cools. Models FT402, FT902 and FT903 include an external Pt100 sensor and display for input of temperature set point. The FT200 has a 9 cm long x 4 cm diameter cooling dumbbell. The FT400 / FT402 have a 12 cm long x 5 cm diameter cooling dumbbell. The FT900 / FT902 have a flexible cooling probe 65 cm long / 1.5 cm diameter.
Catalog No. 50-753-3999 Supplier Julabo USA Inc Supplier No. 9650890.2
Add to Cart
Add to Cart
Request a Quote
Chillers, Julabo, FT900 Immersion Cooler -90¿+30deg.C; 300W cooling power. JULABO Immersion Coolers provide counter-cooling when connected to a heating circulator or rapidly cool fluids down to a low temperature. With easy operation the units are suitable for a variety of cooling applications. Units are an alternative to cooling with dry ice or tap water.The FT200, FT400 and F900 units simply turn on and the probe cools. Models FT402, FT902 and FT903 include an external Pt100 sensor and display for input of temperature set point. The FT200 has a 9 cm long x 4 cm diameter cooling dumbbell. The FT400 / FT402 have a 12 cm long x 5 cm diameter cooling dumbbell. The FT900 / FT902 have a flexible cooling probe 65 cm long / 1.5 cm diameter.