Learn More
Cole-Parmer Fischer Technology 604-142 Thickness Gauge Probe 0 to 45 mils, for models Isoscope FMP10 & Dualscope FMP20
Catalog No. NC0590175 Shop All Cole-Parmer Products
Encompass_Preferred
Accuracy: 0.5 m up to 50 m; 1% from 50 to 800 m; < 3 % above 800 m. Available for sale in the United States, Canada, and Mexico only.
Catalog No. NC0590175 Supplier Cole-Parmer Supplier No. 5410008
Accuracy: 0.5 m up to 50 m; 1% from 50 to 800 m; < 3 % above 800 m. Available for sale in the United States, Canada, and Mexico only.