Learn More
Mopec FTA™ Card
SDP
Catalog No. 22446036 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
1.5 x 3 in.
1.88 x 2.88 in.
3 x 5 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-446-036 1.88 x 2.88 in.
22-446-001 1.5 x 3 in.
22-446-031 3 x 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-446-036 Supplier Mopec Supplier No. FS252
Add to Cart
Add to Cart
Designed for collection, transport, storage, and purification of nucleic acids from biological samples.
  • Each circle holds 100 to 125 μL
  • Features an integral cover sheet to protect the sample.
  • Allows the card to stand on its own while drying.
  • Available in multiple sizes.
  • Features Pre-defined sample areas.
Specifications
Dimensions (L x W) 1.88 x 2.88 in.
Product Type Specimen Collection Card
Color White with Black Print
Quantity 100/Pk.
Style 2 Spots