Learn More
Mopec FTA Indicating Card
SDP
Catalog No. 22446055 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
1.5 x 3 in.
1.88 x 2.88 in.
3 x 5 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-446-055 3 x 5 in.
22-445-983 1.5 x 3 in.
22-446-039 1.88 x 2.88 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-446-055 Supplier Mopec Supplier No. FS256
Add to Cart
Add to Cart
Designed to collect and store buccal cell and saliva samples for DNA analysis.
  • Each circle holds 100 to 125 μL
  • Features an integral cover sheet for sample protection and stability during drying
  • Available in multiple sizes to accommodate different sample capacities.
  • Features a color indicator that changes from pink to white when a sample is applied.
  • For collection and storage of buccal cell and saliva samples for DNA analysis
Specifications
Dimensions (L x W) 3 x 5 in.
Product Type Indicator Card
Quantity 100/Pk.
Style 4 Spots