Learn More
ATCC Verified STR Profiling Service
Catalog No. 502384694
Encompass
FTA Sample Collection Kit for Human Cell Authentication Service utilizes short tandem repeat (STR) profiling. Trained experts amplify 17 STR loci plus Amelogenin using Promega’s PowerPlex18D System. A comprehensive analysis report interprets both karyotypically normal and abnormal cell lines, includes a electropherograms supporting the allele calls at each locus, known reference profiling against the ATCC STR database and a comprehensive interpretation of results. Sample kit mailed upon purchase. Download the instructions for sample preparation and submission.
Catalog No. 50-238-4694 Supplier ATCC Supplier No. 135XV
 Restricted Item
Add to Cart
Add to Cart
FTA Sample Collection Kit for Human Cell Authentication Service utilizes short tandem repeat (STR) profiling. Trained experts amplify 17 STR loci plus Amelogenin using Promega’s PowerPlex18D System. A comprehensive analysis report interprets both karyotypically normal and abnormal cell lines, includes a electropherograms supporting the allele calls at each locus, known reference profiling against the ATCC STR database and a comprehensive interpretation of results. Sample kit mailed upon purchase. Download the instructions for sample preparation and submission.