Learn More
Cayman Chemical FTase InhIbItr I 500ug
SDP
Catalog No. 502995506
Encompass_Preferred
A potent and selective FTase inhibitor (IC50 = 21 nM); greater than 30-fold selective for FTase over GGTase (IC50 = 790 nM); prevents farnesylation of Ras; inhibits proliferation of cells transformed by Ras farnesylation; reduces GTP-Ras, decreases angiographic vasospasm, and improves clinical scores in a dog model of subarachnoid hemorrhage
Catalog No. 50-299-5506 Supplier Cayman Chemical Supplier No. 22755500 UG
A potent and selective FTase inhibitor (IC50 = 21 nM); greater than 30-fold selective for FTase over GGTase (IC50 = 790 nM); prevents farnesylation of Ras; inhibits proliferation of cells transformed by Ras farnesylation; reduces GTP-Ras, decreases angiographic vasospasm, and improves clinical scores in a dog model of subarachnoid hemorrhage