Learn More
Cayman Chemical FTase Inhibitor II (trifluoroacetate salt), Purity: ≥80%, 1 mg
SDP
Catalog No. 502965443
Encompass_Preferred
A cell-permeable analog of FPP that potently inhibits FTase (IC50 = 50-75 nM), preventing farnesylation of Ras; does not inhibit geranylgeranyl transferase at similar concentrations (IC50 > 100 µM); blocks Ras-mediated transformation of NIH 3T3 cells
Catalog No. 50-296-5443 Supplier Cayman Chemical Supplier No. 166071 MG
A cell-permeable analog of FPP that potently inhibits FTase (IC50 = 50-75 nM), preventing farnesylation of Ras; does not inhibit geranylgeranyl transferase at similar concentrations (IC50 > 100 µM); blocks Ras-mediated transformation of NIH 3T3 cells