Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ftaxilide | 19368-18-4 | 98.9% | 100 MG
SDP

Catalog No. 5000412228
Encompass_Preferred

Ftaxilide is a novel antituberculosis agent for research use only. This product features:

  • High purity (98.94%)
  • Molecular weight of 269.30
  • Chemical formula C16H15NO3
  • Solid appearance, white to off-white in color
  • Recommended storage conditions: -20°C for powder up to 3 years, -80°C in solvent up to 2 years
  • Solubility in DMSO: ≥ 40 mg/mL

Catalog No. 50-004-12228 Supplier Medchemexpress LLC Supplier No. HYB1040100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ftaxilide is a novel antituberculosis agent for research use only. This product features:

  • High purity (98.94%)
  • Molecular weight of 269.30
  • Chemical formula C16H15NO3
  • Solid appearance, white to off-white in color
  • Recommended storage conditions: -20°C for powder up to 3 years, -80°C in solvent up to 2 years
  • Solubility in DMSO: ≥ 40 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.