Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ftaxilide | 19368-18-4 | 98.94% | 50 MG
SDP

Catalog No. 5000413103
Encompass_Preferred

Ftaxilide is a novel antituberculosis agent with a molecular weight of 269.30 and a chemical formula of C16H15NO3. It is characterized as a white to off-white solid with a high purity of 98.94%. This compound is intended for research use only and is not available for patient use.

  • Novel antituberculosis agent offers potential for new research in the fight against tuberculosis.
  • High purity of 98.94% ensures reliable results for research applications.
  • Defined chemical properties with molecular weight 269.30, chemical formula C16H15NO3, and CAS number 19368-18-4.
  • Good solubility in DMSO at ≥ 40 mg/mL (148.53 mM), facilitating in vitro experiments.
  • Stable storage as powder for 3 years at -20°C and 2 years at 4°C.
  • Stable storage in solvent for 2 years at -80°C and 1 year at -20°C.
  • Research-grade product, not for direct medical use.

Catalog No. 50-004-13103 Supplier Medchemexpress LLC Supplier No. HYB104050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ftaxilide is a novel antituberculosis agent with a molecular weight of 269.30 and a chemical formula of C16H15NO3. It is characterized as a white to off-white solid with a high purity of 98.94%. This compound is intended for research use only and is not available for patient use.

  • Novel antituberculosis agent offers potential for new research in the fight against tuberculosis.
  • High purity of 98.94% ensures reliable results for research applications.
  • Defined chemical properties with molecular weight 269.30, chemical formula C16H15NO3, and CAS number 19368-18-4.
  • Good solubility in DMSO at ≥ 40 mg/mL (148.53 mM), facilitating in vitro experiments.
  • Stable storage as powder for 3 years at -20°C and 2 years at 4°C.
  • Stable storage in solvent for 2 years at -80°C and 1 year at -20°C.
  • Research-grade product, not for direct medical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.