Learn More
Aviva Systems Biology Corp FTCD Antibody - middle region (ARP41577_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41577_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FTCD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FTCD. Alternate names: LCHC1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57912