Learn More
Thermo Scientific Ferrule Double Cone High Pressure
Supplier:  Thermo Scientific 074373
Encompass
HP double cone shaped ferrule with 0.53 MM inner diameter forms leak-free seal on outside of capillary tubing for connecting to fittings in gas chromatography & other analytical chemistry equipment.
Catalog No. 50-134-5860