Learn More
Aviva Systems Biology Corp FTH1 Antibody - N-terminal region (ARP54619_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54619_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FTH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FTH1. Alternate names: FTH, FTHL6, MGC104426, PIG15, PLIF, FHCApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-62972