Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fti-2153 | 344900-92-1 | 99.8% | 466.60 g/mol | C25H30N4O3S | 100MG
SDP

Catalog No. 5000236606
Encompass_Preferred

FTI-2153 is a potent, highly selective inhibitor of farnesyltransferase (FTase) used as a research compound with reported anti-cancer activity; it inhibits FTase with an IC50 of 1.4 nM and shows strong selectivity for blocking H-Ras processing.

  • Potent FTase inhibition (IC50 1.4 nM).
  • Demonstrated selectivity for H-Ras processing.
  • High chemical purity (99.8%).
  • Molecular formula C25H30N4O3S; molecular weight 466.60 g/mol.
  • Solid physical form suited for laboratory use.
  • Storage under nitrogen at -20°C; in solvent store at -80°C for up to 6 months.

Catalog No. 50-002-36606 Supplier Medchemexpress LLC Supplier No. HY123242100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FTI-2153 is a potent, highly selective inhibitor of farnesyltransferase (FTase) used as a research compound with reported anti-cancer activity; it inhibits FTase with an IC50 of 1.4 nM and shows strong selectivity for blocking H-Ras processing.

  • Potent FTase inhibition (IC50 1.4 nM).
  • Demonstrated selectivity for H-Ras processing.
  • High chemical purity (99.8%).
  • Molecular formula C25H30N4O3S; molecular weight 466.60 g/mol.
  • Solid physical form suited for laboratory use.
  • Storage under nitrogen at -20°C; in solvent store at -80°C for up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.