Learn More
Medchemexpress LLC Fti-2153 | 344900-92-1 | MFCD32215316 | 99.8% | 466.60 | C25H30N4O3S | 50MG
SDP
Catalog No. 5000238266
Encompass_Preferred
FTI-2153 is a potent, highly selective farnesyltransferase (FTase) inhibitor for research use. It inhibits FTase with an IC50 of 1.4 nM and is used to study protein prenylation and FTase-related cellular processes.
Catalog No. 50-002-38266 Supplier Medchemexpress LLC Supplier No. HY12324250MG
FTI-2153 is a potent, highly selective farnesyltransferase (FTase) inhibitor for research use. It inhibits FTase with an IC50 of 1.4 nM and is used to study protein prenylation and FTase-related cellular processes.