Learn More
Medchemexpress LLC FTI-2153 (TFA) | 2820151-01-5 | 99.1% | 580.62 g/mol | C27H31F3N4O5S | 10MM 1ML
SDP
Catalog No. 5000239009
Encompass_Preferred
FTI-2153 TFA is the trifluoroacetic acid salt of a potent farnesyltransferase inhibitor supplied for research use. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and in solid amounts, with supporting datasheet, SDS, and COA for characterization and safe handling.
Catalog No. 50-002-39009 Supplier Medchemexpress LLC Supplier No. HY123242A10MM 1ML
FTI-2153 TFA is the trifluoroacetic acid salt of a potent farnesyltransferase inhibitor supplied for research use. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and in solid amounts, with supporting datasheet, SDS, and COA for characterization and safe handling.