Learn More
Medchemexpress LLC Fti-2153 TFA | 2820151-01-5 | MFCD30187871 | 99.1% | 580.62 | C27H31F3N4O5S | 100MG
SDP
Catalog No. 5000238270
Encompass_Preferred
FTI-2153 TFA is a potent and selective farnesyltransferase inhibitor supplied as the trifluoroacetic acid salt for research use. It inhibits FTase at low nanomolar concentrations, blocks H-Ras processing, and induces mitotic spindle defects and mitotic arrest in cellular studies of Ras signaling and anticancer mechanisms. Available as a high-purity solid or as a ready-to-use DMSO solution.
Catalog No. 50-002-38270 Supplier Medchemexpress LLC Supplier No. HY123242A100MG
FTI-2153 TFA is a potent and selective farnesyltransferase inhibitor supplied as the trifluoroacetic acid salt for research use. It inhibits FTase at low nanomolar concentrations, blocks H-Ras processing, and induces mitotic spindle defects and mitotic arrest in cellular studies of Ras signaling and anticancer mechanisms. Available as a high-purity solid or as a ready-to-use DMSO solution.