Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fti-2153 trifluoroacetate (TFA salt) | 2820151-01-5 | MFCD33548923 | 99.1% | 580.62 | C27H31F3N4O5S | 50MG
SDP

Catalog No. 5000237739
Encompass_Preferred

FTI-2153 TFA is the trifluoroacetic acid salt of a potent, selective farnesyltransferase (FTase) inhibitor provided as a research reagent for biochemical and cell-based studies targeting FTase and oncogenic H-Ras signaling.

  • Trifluoroacetic acid (TFA) salt form for improved handling and solubility.
  • High reported purity of 99.11% for reliable experimental results.
  • Available in small pack sizes suited to research workflows, including 50 mg.
  • Suitable for biochemical and cell-based FTase inhibition assays.
  • Molecular weight 580.62 and formula C27H31F3N4O5S for reference in calculations.

Catalog No. 50-002-37739 Supplier Medchemexpress LLC Supplier No. HY123242A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FTI-2153 TFA is the trifluoroacetic acid salt of a potent, selective farnesyltransferase (FTase) inhibitor provided as a research reagent for biochemical and cell-based studies targeting FTase and oncogenic H-Ras signaling.

  • Trifluoroacetic acid (TFA) salt form for improved handling and solubility.
  • High reported purity of 99.11% for reliable experimental results.
  • Available in small pack sizes suited to research workflows, including 50 mg.
  • Suitable for biochemical and cell-based FTase inhibition assays.
  • Molecular weight 580.62 and formula C27H31F3N4O5S for reference in calculations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.