Learn More
Medchemexpress LLC Fti-276 trifluoroacetate | 1217471-51-6 | 98.0% | 547.6 g/mol | C23H28F3N3O5S2 | 10 MG
SDP
Catalog No. 5000287973
Encompass_Preferred
FTI-276 trifluoroacetate is the trifluoroacetate salt of a small-molecule inhibitor of protein farnesyl transferase. It is provided as a solid for research use and exhibits sub-nanomolar activity against both Plasmodium falciparum and human PFT, making it suitable for biochemical and antiparasitic research applications.
  • Potent protein farnesyl transferase inhibition at sub-nanomolar concentrations.
  • Provided as the trifluoroacetate (TFA) salt to aid handling and formulation.
  • High reported purity, approximately 98%.
  • Supplied in small, research-ready quantities suitable for assay development.
  • Characterized by published chemical identifiers for unambiguous compound identification.
Catalog No. 50-002-87973 Supplier Medchemexpress LLC Supplier No. HY15873A10MG
FTI-276 trifluoroacetate is the trifluoroacetate salt of a small-molecule inhibitor of protein farnesyl transferase. It is provided as a solid for research use and exhibits sub-nanomolar activity against both Plasmodium falciparum and human PFT, making it suitable for biochemical and antiparasitic research applications.
  • Potent protein farnesyl transferase inhibition at sub-nanomolar concentrations.
  • Provided as the trifluoroacetate (TFA) salt to aid handling and formulation.
  • High reported purity, approximately 98%.
  • Supplied in small, research-ready quantities suitable for assay development.
  • Characterized by published chemical identifiers for unambiguous compound identification.