Learn More
Apexbio Technology LLC FTI 277 HCl, 50mg Cas# 180977-34-8
SDP
Supplier:  Apexbio Technology LLC B584250
Encompass_Preferred
FTase inhibitor. FTI 277 HCl 5mg. 1346572-63-1. MFCD28405142 FTI-277 is a highly potent and selective FTase inhibitor. At concentration as low as 10 nM, FTI-277 inhibits H-Ras processing and the processing is blocked by more than 95% at 3 M. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8715