Learn More
Selleck Chemical LLC FTI 277 HCl 5mg 180977-34-8
SDP
Catalog No. 501364818
Encompass_Preferred
FTI 277 HCl is the methyl ester of FTI 277, which is a potent and selective farnesyltransferase (FTase) inhibitor with IC50 of 500 pM, about 100-fold selectivity over the closely related GGTase I. FTI 277 HCl inhibits cell growth and induces apoptosis. FTI 277 HCl is effective in clearing HDV viremia. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4818 Supplier Selleck Chemical LLC Supplier No. S74655MG
FTI 277 HCl is the methyl ester of FTI 277, which is a potent and selective farnesyltransferase (FTase) inhibitor with IC50 of 500 pM, about 100-fold selectivity over the closely related GGTase I. FTI 277 HCl inhibits cell growth and induces apoptosis. FTI 277 HCl is effective in clearing HDV viremia. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.