Learn More
Selleck Chemical LLC FTI 277 HCl
SDP
Catalog No. 5000004438
Encompass_Preferred
FTI 277 HCl is a potent and selective farnesyltransferase (FTase) inhibitor with IC50 of 500 pM about 100-fold selectivity over the closely related GGTase I FTI 277 HCl inhibits cell growth and induces apoptosis FTI 277 HCl is effective in clearing HDV viremia
Catalog No. 50-000-04438 Supplier Selleck Chemical LLC Supplier No. S746510MM/1ML
Add to Cart
Add to Cart
FTI 277 HCl is a potent and selective farnesyltransferase (FTase) inhibitor with IC50 of 500 pM about 100-fold selectivity over the closely related GGTase I FTI 277 HCl inhibits cell growth and induces apoptosis FTI 277 HCl is effective in clearing HDV viremia