Learn More
TARGETMOL CHEMICALS INC FTI-277 HYDROCHLORIDE 10MG
SDP
Catalog No. 502580657
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FTI-277 hydrochloride (FTI 277 HCl) is an effective and specific farnesyltransferase (FTase) inhibitor (IC50 500 pM); this selectivity is about 100-fold than the closely related GGTase I. purity: 98%
Catalog No. 50-258-0657 Supplier TARGETMOL CHEMICALS INC Supplier No. T270010MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FTI-277 hydrochloride (FTI 277 HCl) is an effective and specific farnesyltransferase (FTase) inhibitor (IC50 500 pM); this selectivity is about 100-fold than the closely related GGTase I. purity: 98%