Learn More
Medchemexpress LLC FTI-277 hydrochloride | 180977-34-8 | 98.0% | 484.07 | 5 MG
SDP
Catalog No. 5000364660
Encompass_Preferred
FTI-277 hydrochloride is an inhibitor of farnesyl transferase (FTase). It is a highly potent Ras CAAX peptidomimetic that antagonizes both H- and K-Ras oncogenic signaling and can inhibit hepatitis delta virus (HDV) infection.
  • Inhibits farnesyl transferase (FTase)
  • Antagonizes H- and K-Ras oncogenic signaling
  • Can inhibit hepatitis delta virus (HDV) infection
  • Significantly decreases survival of radioresistant cells expressing the 24-kDa isoform
  • Stimulates postmitotic cell death in HeLa 3A cells
  • Reduces G(2)/M-phase arrest in both cell types (HeLa 3A and HeLa PINA)
  • Inhibits migration and invasion in PC-3 cells
  • Prevents increased PTP-1B and PTEN protein expression in burned mice
Catalog No. 50-003-64660 Supplier Medchemexpress LLC Supplier No. 5MGHY15872A
Add to Cart
Add to Cart
FTI-277 hydrochloride is an inhibitor of farnesyl transferase (FTase). It is a highly potent Ras CAAX peptidomimetic that antagonizes both H- and K-Ras oncogenic signaling and can inhibit hepatitis delta virus (HDV) infection.
  • Inhibits farnesyl transferase (FTase)
  • Antagonizes H- and K-Ras oncogenic signaling
  • Can inhibit hepatitis delta virus (HDV) infection
  • Significantly decreases survival of radioresistant cells expressing the 24-kDa isoform
  • Stimulates postmitotic cell death in HeLa 3A cells
  • Reduces G(2)/M-phase arrest in both cell types (HeLa 3A and HeLa PINA)
  • Inhibits migration and invasion in PC-3 cells
  • Prevents increased PTP-1B and PTEN protein expression in burned mice