Learn More
SCP Science Conostan FTIR BR 1.0 Test Standard, 100 g
Catalog No. NC1823992
Encompass
Our FTIR operational test standards is a petroleum oil-based fluid that looks and handles like routinely tested used-oil samples. It is designed for validating FTIR instrument performance in order to ensure repeatability and reproducibility.
Catalog No. NC1823992 Supplier SCP Science Supplier No. 150702001
Our FTIR operational test standards is a petroleum oil-based fluid that looks and handles like routinely tested used-oil samples. It is designed for validating FTIR instrument performance in order to ensure repeatability and reproducibility.