Learn More
TA Instruments FTIR Gas Cell iS5 iS10, Heated FTIR Gas Cell, KBr Windows, Electronic Temperature Controller up to 350°C, Zero Gravity Principle, 100 mm Optical Path, 11 ml Internal Volume, Sellable Only with Separately Sold EGA Gas Sampling Device
Catalog No. 5000580843 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments FTIR Gas Cell for EGA with Thermo Nicolet iS5 or iS10 FTIR is used with a separately sold EGA Gas Sampling Device. The heated cell includes KBr windows and an electronic temperature controller up to 350 °C. It is based on the Zero Gravity principle with a 100 mm optical path and 11 ml internal volume. It is sellable and functional only with a separately sold EGA Gas Sampling Device.
Catalog No. 50-005-80843 Supplier TA Instruments Supplier No. 206412.002
Waters TA Instruments FTIR Gas Cell for EGA with Thermo Nicolet iS5 or iS10 FTIR is used with a separately sold EGA Gas Sampling Device. The heated cell includes KBr windows and an electronic temperature controller up to 350 °C. It is based on the Zero Gravity principle with a 100 mm optical path and 11 ml internal volume. It is sellable and functional only with a separately sold EGA Gas Sampling Device.