Learn More
Perkin Elmer US LLC FT-IR Pharmacopeia Compliance Resource Pack
Catalog No. 502117244
Encompass_Preferred
The documentation provided in the pack is structured to mirror the steps in the analysis process; namely: FT-IR Instrument Performance Verification FT-IR Sampling Methods and Pharmacopeia best practice FT-IR Pharmacopeia Identification Tests 21 CFR Part 11 Technical Compliance
Catalog No. 50-211-7244 Supplier Perkin Elmer US LLC Supplier No. L1608004
The documentation provided in the pack is structured to mirror the steps in the analysis process; namely: FT-IR Instrument Performance Verification FT-IR Sampling Methods and Pharmacopeia best practice FT-IR Pharmacopeia Identification Tests 21 CFR Part 11 Technical Compliance