Learn More
Perkin Elmer US LLC FT-IR Polymer Resource Pack
Catalog No. 502117242
Encompass_Preferred
In todays competitive polymer market, making efficient use of time and resources is vital. The Spectrum Two with the FT-IR Polymer QA/QC Resource Pack can be set up in minutes to deliver the answer you need, fast. This Polymer QA/QC Resource Pack includes: A guide encompassing a description of the applications of infrared spectroscopy and ATR for identification of polymers A starter mid-IR spectral library of various reference polymers The Polymer Resource Pack is designed to help in the following: Measurement of good quality spectrum of polymers Identification of polymers Interpretation of polymer spectra Utilization of library spectra is illustrated in this resource pack for effective results. The documentation provides clear instructions for day-to-day use of the system and demonstrates the value of using UATR-FT-IR spectroscopy in the identification and verification of materials.
Catalog No. 50-211-7242 Supplier Perkin Elmer US LLC Supplier No. L1608013
In todays competitive polymer market, making efficient use of time and resources is vital. The Spectrum Two with the FT-IR Polymer QA/QC Resource Pack can be set up in minutes to deliver the answer you need, fast. This Polymer QA/QC Resource Pack includes: A guide encompassing a description of the applications of infrared spectroscopy and ATR for identification of polymers A starter mid-IR spectral library of various reference polymers The Polymer Resource Pack is designed to help in the following: Measurement of good quality spectrum of polymers Identification of polymers Interpretation of polymer spectra Utilization of library spectra is illustrated in this resource pack for effective results. The documentation provides clear instructions for day-to-day use of the system and demonstrates the value of using UATR-FT-IR spectroscopy in the identification and verification of materials.