Learn More
Medchemexpress LLC FTISADTSK-13C6,15N2(TFA) | 96.04% | 1091.01 | 5 MG
SDP
Catalog No. 5000397467
Encompass_Preferred
FTISADTSK-13C6,15N2 TFA is a 13C- and 15N-labeled stable isotope peptide derived from Trastuzumab. It is an endogenous stable signature peptide monitored by selected reaction monitoring (SRM) and is used as a tracer for quantitation during drug development.
  • 13C- and 15N-labeled stable isotope peptide
  • Derived from Trastuzumab
  • Monitored by selected reaction monitoring (SRM)
  • Used as a tracer for quantitation during drug development
Catalog No. 50-003-97467 Supplier Medchemexpress LLC Supplier No. HYP3146S5MG
FTISADTSK-13C6,15N2 TFA is a 13C- and 15N-labeled stable isotope peptide derived from Trastuzumab. It is an endogenous stable signature peptide monitored by selected reaction monitoring (SRM) and is used as a tracer for quantitation during drug development.
  • 13C- and 15N-labeled stable isotope peptide
  • Derived from Trastuzumab
  • Monitored by selected reaction monitoring (SRM)
  • Used as a tracer for quantitation during drug development