Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2512MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: FTL/1388. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a. Positive Control: HepG2, HeLa, HL-60 or 293T cells. Pancreas, Liver, Cerebellum or Testis. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human FTL protein fragment (aa 38-165). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 19-25kDa.Chromosome: 19q13.33. Gene Symbol: FTL. Gene ID: 2512. Ensemble: ENSG00000087086. Uniprot: NP_000137.
Catalog No. 50-188-4720