Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ FTM Double Bag (100 mL Septum)

Catalog No. R112976
Change view
Click to view available options
100 mL Septum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
R112976 100 mL Septum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R112976 Supplier Thermo Scientific™ Supplier No. R112976
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ Fluid Thioglycollate media meets or exceeds USP growth promotion requirments.

  • Fluid Thioglycollate media was tested according to USP growth promotion testing procedure.
  • Inoculation levels of 10 to 100 CFUs were used, and incubation was conducted at 30 to 35°C for not more than 72 hours.
  • The results meet or exceed USP growth promotion requirements.
TRUSTED_SUSTAINABILITY

Specifications

Broth Type General growth media
Color Amber
Content And Storage 2°C to 8°C
Format Double bagged bottle
Media Type Culture Media
Fill Volume 100 mL
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.