Learn More
Alkali Scientific FTM Formulation 10mL 16x125 (Fluid Thicoglycllate) 16x125mm, 10/Pk
SDP
Catalog No. 5000662024
Encompass_Preferred
Fluid Thioglycollate Medium (FTM), 10mL Tubes, 10/Pk is a ready to use sterility testing medium formulated for the detection of aerobic and anaerobic microorganisms in pharmaceutical, medical device, and biotechnology applications. The enriched formulation supports growth of a wide range of microorganisms and is widely used for sterility validation and microbial monitoring. Sodium thioglycollate acts as a reducing agent to create suitable conditions for anaerobic growth, while resazurin functions as a redox indicator to monitor oxygen levels in the medium. Supplied in sterile 10mL tubes, the format allows direct inoculation or swab immersion during sterility testing procedures. FTM supports reliable detection of aerobic, anaerobic, and facultative organisms during routine microbiological quality control testing.
Catalog No. 50-006-62024 Supplier Alkali Scientific Supplier No. FTMT00
Fluid Thioglycollate Medium (FTM), 10mL Tubes, 10/Pk is a ready to use sterility testing medium formulated for the detection of aerobic and anaerobic microorganisms in pharmaceutical, medical device, and biotechnology applications. The enriched formulation supports growth of a wide range of microorganisms and is widely used for sterility validation and microbial monitoring. Sodium thioglycollate acts as a reducing agent to create suitable conditions for anaerobic growth, while resazurin functions as a redox indicator to monitor oxygen levels in the medium. Supplied in sterile 10mL tubes, the format allows direct inoculation or swab immersion during sterility testing procedures. FTM supports reliable detection of aerobic, anaerobic, and facultative organisms during routine microbiological quality control testing.