Learn More
Biomatik Corporation Ftna Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11913
Encompass_Preferred
Ftna Polyclonal Antibody is a Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) Polyclonal Antibody to ftnA. The Ftna Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori). Immunogen: Recombinant Helicobacter Pylori Bacterial Non-Heme Ferritin (Ftna) (1-167Aa). Applications: ELISA, WB. Target Synonyms: ftnA. Accession Number: P52093. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5376