Learn More
MyBioSource ftnA Recombinant Protein
SDP
Supplier:  MyBioSource MBS1572840.1 MG
Encompass_Preferred
The ftnA Recombinant Protein is a recombinant protein from Francisella tularensis. It is produced in E. coli and has a molecular weight of approximately 17.5 kDa. This protein can be used in activity assays, ELISA, Western blots, and other analytical applications. It is supplied as a lyophilized powder and should be reconstituted in water. The recommended concentration for optimal use is 0.2-0.5 mg/mL. This product can enable research on the biological functions and interactions of this Francisella tularensis protein.
Catalog No. NC2199088