Learn More
AVANTOR MASTERFLEX (VWR LLC) FTNG ELBOW 1/4FTFTX3/16FTFT 10
Catalog No. 502742288
Encompass
Masterflex Fitting, Polypropylene, Elbow, Hosebarb to Thread, 3/16IN ID x 1/4IN NPT(M)10/PK, Avantor
Catalog No. 50-274-2288 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4062295
Masterflex Fitting, Polypropylene, Elbow, Hosebarb to Thread, 3/16IN ID x 1/4IN NPT(M)10/PK, Avantor